Tisk…

Dysprosium(III) acetate tetrahydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Dodavatel: Thermo Scientific

11314.09 11314.18
11314.09EA 2200 CZK
11314.09 11314.18
Dysprosium(III) acetate tetrahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Dysprosium(III) acetate tetrahydrate

Purity: 99.99% CAS No.: 15280-55-4 Molecular Formula: Dy(OOCCH3)3.4H2O Formula Weight: 411.68 (339.62anhy) Physical Form: Crystalline Sensitivity: Hygroscopic MDL No.: MFCD00013033

Vzorec: C₆H₉DyO₆·4H₂O
Bod tání: 120 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00013033
CAS číslo: 15280-55-4
EINECS: 242-565-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR