Tisk…

Benzyl-4-chlorophenyl ketone ≥98%

Dodavatel: Thermo Scientific

Synonyms: Benzyl 4-chlorophenyl ketone, 4'-Chloro-2-phenylacetophenone

A15959.06 A15959.14 A15959.22
A15959.06EA 1550 CZK
A15959.06 A15959.14 A15959.22
Benzyl-4-chlorophenyl ketone ≥98%
Benzyl-4-chlorophenyl ketone

Purity: 98% CAS No.: 1889-71-0 Molecular Formula: C14H11ClO Formula Weight: 230.69 Melting Point: 103-107° Beilstein Reference: 976443

Vzorec: C₁₄H₁₁ClO
MW: 230,69 g/mol
Bod tání: 103…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00016342
CAS číslo: 1889-71-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR