Tisk…

Dioctyldichlorosilane ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

42599.09 42599.18
42599.09EA 2810 CZK
42599.09 42599.18
Dioctyldichlorosilane ≥97%
Dioctyldichlorosilane

Purity: 97% CAS No.: 18416-07-4 Molecular Formula: C16H34Cl2Si Formula Weight: 325.44 Physical Form: Liquid Density: 0.940 Appearance: Colourless to pale yellow Boiling Point: 145°/0.2mm UN No.: 2987 MDL No.: MFCD01074472

Vzorec: C₁₆H₃₄Cl₂Si
Bod varu: 145 °C (0,2 mmHg)
Hustota: 0,940
Storage Temperature: Ambient
MDL Number: MFCD01074472
CAS číslo: 18416-07-4
UN: 2987
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR