Dichloromethyloctylsilane ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
L16565.09
L16565.18
L16565.09EA
673
CZK
L16565.09
L16565.18
Dichloromethyloctylsilane ≥98%
Dichloromethyloctylsilane
Purity: 98%
CAS No.: 14799-93-0
Molecular Formula: C9H20Cl2Si
Formula Weight: 227.25
Flash Point: 98°(208°F)
Refractive Index: 1.4435
Boiling Point: 224-226°
Beilstein Reference: 1740697
UN No.: 2987
MDL No.: MFCD00042606
Vzorec:
C₉H₂₀Cl₂Si Bod varu: 224…226 °C Hustota: 0,794 Bod vzplanutí: 98 °C (208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042606 CAS číslo: 14799-93-0 EINECS: 238-863-2 UN: 2987 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...