Tisk…

Benzophenone-[D₁₀] (99% D), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

471178-1G 471178-5G
SIAL471178-1GEA 11930 CZK
SIAL471178-1G SIAL471178-5G
Benzophenone-[D₁₀] (99% D), Sigma-Aldrich®
Benzophenone-[D₁₀]
Vzorec: C₁₃D₁₀O
CAS číslo: 22583-75-1
EINECS: 245-107-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR