Geranyl acetate (mixture of isomers), Sigma-Aldrich®
Dodavatel: Merck
45897-100ML-F
45897-500ML-F
SIAL45897-100ML-FEA
3610
CZK
SIAL45897-100ML-F
SIAL45897-500ML-F
Geranyl acetate (mixture of isomers), Sigma-Aldrich®
Geranyl acetate (mixture of isomers)
Vzorec:
C₁₂H₂₀O₂ |
MDL Number:
MFCD00015037 CAS číslo: 16409-44-2 EINECS: 240-458-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...