2,3,3-Trimethylindolenine ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
B22410.06
B22410.14
B22410.06EA
1090
CZK
B22410.06
B22410.14
2,3,3-Trimethylindolenine ≥98%
2,3,3-Trimethylindolenine
Purity: 98%
CAS No.: 1640-39-7
Molecular Formula: C11H13N
Formula Weight: 159.23
Flash Point: 93°(199°F)
Refractive Index: 1.5470
Melting Point: 6-8°
Boiling Point: 228-229°
Beilstein Reference: 119740
MDL No.: MFCD00005724
Vzorec:
C₁₁H₁₃N MW: 159,23 g/mol Bod varu: 228…229 °C Bod tání: 6…8 °C Hustota: 0,992 Bod vzplanutí: 93 °C (199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005724 CAS číslo: 1640-39-7 EINECS: 216-685-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...