Tisk…

6-Aminonicotinamide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 6-Aminopyridine-3-carboxamide

M-Clarity™ quality level = MQ100

A68203-1G A68203-5G
SIALA68203-1GEA 3580 CZK
SIALA68203-1G SIALA68203-5G
6-Aminonicotinamide, Sigma-Aldrich®
6-Aminonicotinamide
Vzorec: C₆H₇N₃O
MDL Number: MFCD00006327
CAS číslo: 329-89-5
EINECS: 206-349-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR