6-Aminonicotinamide
Dodavatel: Cayman Chemical
Synonyms:
6-Aminopyridine-3-carboxamide
10009315-100
10009315-250
10009315-500
10009315-1
CAYM10009315-100EA
613
CZK
CAYM10009315-100
CAYM10009315-250
CAYM10009315-500
CAYM10009315-1
6-Aminonicotinamide
6-Aminonicotinamide
Vzorec:
C₆H₇N₃O MW: 137.14 g/mol |
MDL Number:
MFCD00006327 CAS číslo: 329-89-5 EINECS: 206-349-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...