Tisk…

N,N-Diethylnicotinamide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

D98807-25G D98807-500G
SIALD98807-25GEA 1670 CZK
SIALD98807-25G SIALD98807-500G
N,N-Diethylnicotinamide, Sigma-Aldrich®
N,N-Diethylnicotinamide
Vzorec: C₁₀H₁₄N₂O
MDL Number: MFCD00006386
CAS číslo: 59-26-7
EINECS: 200-418-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR