Copper(II) isopropoxide ≥98% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
35821.06
35821.14
35821.03
35821.06EA
8270
CZK
35821.06
35821.14
35821.03
Copper(II) isopropoxide ≥98% (metals basis)
Copper(II) isopropoxide
Purity: 98%
CAS No.: 23578-23-6
Molecular Formula: [(CH3)2CHO]2Cu
Formula Weight: 181.73
Physical Form: Powder
Sensitivity: Moisture Sensitive
UN No.: 3180
MDL No.: MFCD00074982
Vzorec:
C₆H₁₄CuO₂ MW: 181.73 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00074982 CAS číslo: 23578-23-6 UN: 3181 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...