Tisk…

Diphenyl phosphate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Diphenyl hydrogen phosphate

850608-5G 850608-25G
SIAL850608-5GEA 2360 CZK
SIAL850608-5G SIAL850608-25G
Diphenyl phosphate, Sigma-Aldrich®
Diphenyl phosphate
Vzorec: C₁₂H₁₁O₄P
MW: 250,19 g/mol
MDL Number: MFCD00003033
CAS číslo: 838-85-7
EINECS: 212-657-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR