Diphenyl phosphate 99%
Dodavatel: Thermo Scientific
Synonyms:
Diphenyl hydrogen phosphate
225690050
225690250
ACRO225690050EA
2120
CZK
ACRO225690050
ACRO225690250
Diphenyl phosphate 99%
Diphenyl phosphate
Vzorec:
C₁₂H₁₁O₄P MW: 250,19 g/mol Bod tání: 63…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003033 CAS číslo: 838-85-7 EINECS: 212-657-2 |
Výsledky specifikačního testu
Appearance | White Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 62 to 70 °C |
Titration with NaOH | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...