Tisk…

Thallium(I) hexafluorophosphate ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Thallium(I) hexafluorophosphate(V)

39627.06 39627.14
39627.06EA 9200 CZK
39627.06 39627.14
Thallium(I) hexafluorophosphate ≥97%
Thallium(I) hexafluorophosphate

Purity: 97% min CAS No.: 60969-19-9 Molecular Formula: TlPF6 Formula Weight: 349.34 Physical Form: Crystalline UN No.: 1707 MDL No.: MFCD00049807

Vzorec: F₆PTl
Storage Temperature: Ambient
MDL Number: MFCD00049807
CAS číslo: 60969-19-9
UN: 1707
ADR: 6.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR