Tisk…

4-Methoxybenzoic anhydride ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

L06180.06 L06180.14
L06180.06EA 2360 CZK
L06180.06 L06180.14
4-Methoxybenzoic anhydride ≥98%
4-Methoxybenzoic anhydride

Purity: 98% CAS No.: 794-94-5 Molecular Formula: C16H14O5 Formula Weight: 286.29 Melting Point: 95-99° Beilstein Reference: 545712

Vzorec: C₁₆H₁₄O₅
MW: 286.29 g/mol
Bod tání: 95…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00017175
CAS číslo: 794-94-5
EINECS: 212-345-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR