Tisk…

4-(4-Fluorobenzyloxy)benzaldehyde ≥97%

Dodavatel: Thermo Fisher Scientific

Synonyms: 4-((4-Fluorobenzyl)oxy)benzaldehyde

A17657.06 A17657.14
A17657.06EA 1330 CZK
A17657.06 A17657.14
4-(4-Fluorobenzyloxy)benzaldehyde ≥97%
4-(4-Fluorobenzyloxy)benzaldehyde

Purity: 97% CAS No.: 56442-17-2 Molecular Formula: C14H11FO2 Formula Weight: 230.24 MDL No.: MFCD00173650

Vzorec: C₁₄H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD00173650
CAS číslo: 56442-17-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR