4'-Bromo-2'-hydroxy-5'-methylacetophenone ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-(4-Bromo-2-hydroxy-5-methylphenyl)ethanone
H29125.06
H29125.03
H29125.06EA
2490
CZK
H29125.06
H29125.03
4'-Bromo-2'-hydroxy-5'-methylacetophenone ≥97%
4'-Bromo-2'-hydroxy-5'-methylacetophenone
Purity: 97%
CAS No.: 50342-17-1
Molecular Formula: C9H9BrO2
Formula Weight: 229.07
Melting Point: 86-89°
MDL No.: MFCD08273792
Vzorec:
C₉H₉BrO₂ MW: 229,07 g/mol Bod tání: 86…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD08273792 CAS číslo: 50342-17-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...