Tisk…

4,4'-Diphenoxybenzophenone ≥98%

Dodavatel: Thermo Scientific

Synonyms: Bis(4-phenoxyphenyl)methanone

L13419.03 L13419.06 L13419.14
L13419.03EA 934 CZK
L13419.03 L13419.06 L13419.14
4,4'-Diphenoxybenzophenone ≥98%
4,4'-Diphenoxybenzophenone

Purity: 98% CAS No.: 14984-21-5 Molecular Formula: C25H18O3 Formula Weight: 366.42 Melting Point: 147-151° Beilstein Reference: 2165449

Vzorec: C₂₅H₁₈O₃
MW: 366,42 g/mol
Bod tání: 147…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00077969
CAS číslo: 14984-21-5
EINECS: 403-390-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR