Tisk…

2,2'-Dihydroxy-4-methoxybenzophenone, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

323578-100G 323578-25G
SIAL323578-100GEA 11660 CZK
SIAL323578-100G SIAL323578-25G
2,2'-Dihydroxy-4-methoxybenzophenone, Sigma-Aldrich®
2,2'-Dihydroxy-4-methoxybenzophenone
Vzorec: C₁₄H₁₂O₄
MDL Number: MFCD00002218
CAS číslo: 131-53-3
EINECS: 205-026-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR