Tisk…

4-Chloro-3-nitrobenzophenone ≥99%

Dodavatel: Thermo Scientific
Varování

B23451.14 B23451.22
B23451.14EA 1180 CZK
B23451.14 B23451.22
4-Chloro-3-nitrobenzophenone ≥99%
4-Chloro-3-nitrobenzophenone

Purity: 99% CAS No.: 56107-02-9 Molecular Formula: C13H8ClNO3 Formula Weight: 261.66 Melting Point: 104-106° Boiling Point: 233-235°/12mm MDL No.: MFCD00007081

Vzorec: C₁₃H₈ClNO₃
MW: 261,66 g/mol
Bod varu: 233…235 °C (12 mmHg)
Bod tání: 104…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00007081
CAS číslo: 56107-02-9
EINECS: 259-995-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR