Tisk…

N-Benzylaminoacetaldehyde diethyl acetal 96%

Dodavatel: Thermo Fisher Scientific
Varování

A13841.06 A13841.14
A13841.06EA 1580 CZK
A13841.06 A13841.14
N-Benzylaminoacetaldehyde diethyl acetal 96%
N-Benzylaminoacetaldehyde diethyl acetal

Purity: 96% CAS No.: 61190-10-1 Molecular Formula: C6H5CH2NHCH2CH(OCH2CH3)2 Formula Weight: 223.32 Refractive Index: 1.4890 Boiling Point: 115-117°/1mm MDL No.: MFCD00051599

Vzorec: C₁₃H₂₁NO₂
MW: 223.32 g/mol
Bod varu: 115…117 °C (1 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00051599
CAS číslo: 61190-10-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR