Methyl-4-acetylbenzoate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
544302-25G
544302-5G
SIAL544302-25GEA
9550
CZK
SIAL544302-25G
SIAL544302-5G
Methyl-4-acetylbenzoate, Sigma-Aldrich®
Methyl-4-acetylbenzoate
Vzorec:
C₁₀H₁₀O₃ |
MDL Number:
MFCD00216474 CAS číslo: 3609-53-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...