Tisk…

4,4'-Dimethoxydiphenylamine 98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Bis(4-methoxyphenyl)amine

L01322.03 L01322.06
L01322.03EA 1840 CZK
L01322.03 L01322.06
4,4'-Dimethoxydiphenylamine 98%
4,4'-Dimethoxydiphenylamine

Purity: 98% CAS No.: 101-70-2 Molecular Formula: C14H15NO2 Formula Weight: 229.28 Melting Point: 100-104° Beilstein Reference: 2214262

Vzorec: C₁₄H₁₅NO₂
Bod tání: 100…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00014895
CAS číslo: 101-70-2
EINECS: 202-968-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR