Tisk…

4-(N,N-Diphenylamino)phenylboronic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

647292-1G 647292-5G
SIAL647292-1GEA 5910 CZK
SIAL647292-1G SIAL647292-5G
4-(N,N-Diphenylamino)phenylboronic acid, Sigma-Aldrich®
4-(N,N-Diphenylamino)phenylboronic acid
Vzorec: C₁₈H₁₆BNO₂
MDL Number: MFCD06798117
CAS číslo: 201802-67-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR