Tisk…

Dysprosium(III) isopropoxide 5% (w/v) in 2-propanol/toluene , 99.9% (REO, rare earth oxide basis)

Dodavatel: Thermo Scientific
Nebezpečí

36585.AC 36585.AE
36585.ACEA 5740 CZK
36585.AC 36585.AE
Dysprosium(III) isopropoxide 5% (w/v) in 2-propanol/toluene , 99.9% (REO, rare earth oxide basis)
Dysprosium(III) isopropoxide

Purity: 99.9% CAS No.: 6742-68-3 Molecular Formula: C9H21DyO3 Formula Weight: 339.76 Physical Form: Liquid Sensitivity: Moisture Sensitive Appearance: Brown UN No.: 1993 MDL No.: MFCD00049784

Vzorec: C₉H₂₁DyO₃
MW: 339,761 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00049784
CAS číslo: 6742-68-3
UN: 1993
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR