Tisk…

Tris[N,N-bis(trimethylsilyl)amide]erbium(III) ≥98%

Dodavatel: Thermo Scientific
Nebezpečí


Packaged under argon.

44421.03 44421.06
44421.03EA 4020 CZK
44421.03 44421.06
Tris[N,N-bis(trimethylsilyl)amide]erbium(III) ≥98%
Tris[N,N-bis(trimethylsilyl)amide]erbium(III)

Purity: 98% CAS No.: 103457-72-3 Molecular Formula: C18H54ErN3Si6 Formula Weight: 648.41 Physical Form: Powder- Packaged under argon Sensitivity: Moisture Sensitive Appearance: Pink Melting Point: 166° UN No.: 3396 MDL No.: MFCD00271002

Vzorec: C₁₈H₅₄ErN₃Si₆
MW: 648,41 g/mol
Bod tání: 166 °C
Storage Temperature: Ambient
MDL Number: MFCD00271002
CAS číslo: 103457-72-3
UN: 3396
ADR: 4.3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR