Tisk…

3-Amino-4-bromo-5-phenyl-1H-pyrazole 96%

Dodavatel: Thermo Fisher Scientific
Varování

A10621.03 A10621.06
A10621.03EA 800 CZK
A10621.03 A10621.06
3-Amino-4-bromo-5-phenyl-1H-pyrazole 96%
3-Amino-4-bromo-5-phenyl-1H-pyrazole

Purity: 96% CAS No.: 2845-78-5 Molecular Formula: C9H8BrN3 Formula Weight: 238.09 Odour: Odourless Melting Point: 111-117° MDL No.: MFCD00082666

Vzorec: C₉H₈BrN₃
Bod tání: 111…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00082666
CAS číslo: 2845-78-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR