4-Iodo-1-methyl-1H-pyrazole, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
683531-1G
683531-5G
SIAL683531-1GEA
0
CZK
SIAL683531-1G
SIAL683531-5G
4-Iodo-1-methyl-1H-pyrazole, Sigma-Aldrich®
4-Iodo-1-methyl-1H-pyrazole
Vzorec:
C₄H₅IN₂ MW: 208,0000 g/mol |
MDL Number:
MFCD05663860 CAS číslo: 39806-90-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...