2,3-Dimethylmaleic anhydride ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
3,4-Dimethylfuran-2,5-dione
L12919.06
L12919.14
L12919.06EA
2330
CZK
L12919.06
L12919.14
2,3-Dimethylmaleic anhydride ≥97%
2,3-Dimethylmaleic anhydride
Purity: 97%
CAS No.: 766-39-2
Molecular Formula: C6H6O3
Formula Weight: 126.11
Melting Point: 93-96°
Boiling Point: 222-223°
MDL No.: MFCD00005523
Vzorec:
C₆H₆O₃ MW: 126,11 g/mol Bod varu: 222…223 °C Bod tání: 93…96 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005523 CAS číslo: 766-39-2 EINECS: 212-165-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...