3,4-(Methylenedioxy)phenylboronic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
B24217.03
B24217.06
B24217.14
B24217.03EA
1780
CZK
B24217.03
B24217.06
B24217.14
3,4-(Methylenedioxy)phenylboronic acid ≥98%
3,4-(Methylenedioxy)phenylboronic acid
Purity: 98%
CAS No.: 94839-07-3
Molecular Formula: C7H7BO4
Formula Weight: 165.94
Melting Point: 239-243°
Beilstein Reference: 5523347
Vzorec:
C₇H₇BO₄ Bod tání: 239…243 °C Storage Temperature: Ambient |
MDL Number:
MFCD01009695 CAS číslo: 94839-07-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...