Tisk…

Phthalimidoacetaldehyde diethyl acetal ≥99%

Dodavatel: Thermo Scientific

Synonyms: 2-(2,2-Diethoxyethyl)isoindoline-1,3-dione, Phthalimidoacetaldehyd Diethylacetal

A18491.06 A18491.14
A18491.06EA 1260 CZK
A18491.06 A18491.14
Phthalimidoacetaldehyde diethyl acetal ≥99%
Phthalimidoacetaldehyde diethyl acetal

Purity: 99% CAS No.: 78902-09-7 Molecular Formula: C14H17NO4 Formula Weight: 263.30 Melting Point: 71-74° Beilstein Reference: 215793

Vzorec: C₁₄H₁₇NO₄
MW: 263,30 g/mol
Bod tání: 71…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00005901
CAS číslo: 78902-09-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR