Tisk…

1-Boc-4-(hydroxymethyl)piperidine ≥97%

Dodavatel: Thermo Scientific

Synonyms: tert-butyl 4-(hydroxymethyl)piperidine-1-carboxylate, 1-Boc-4-piperidinemethanol, N-BOC-4-Piperidinemethanol

H26650.03 H26650.06
H26650.03EA 1180 CZK
H26650.03 H26650.06
1-Boc-4-(hydroxymethyl)piperidine ≥97%
1-Boc-4-(hydroxymethyl)piperidine

Purity: 97% CAS No.: 123855-51-6 Molecular Formula: C11H21NO3 Formula Weight: 215.29 Melting Point: 78-82° MDL No.: MFCD02094488

Vzorec: C₁₁H₂₁NO₃
MW: 215,29 g/mol
Bod tání: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD02094488
CAS číslo: 123855-51-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR