Tisk…

9,12-Octadecadiynoic acid

Dodavatel: Cayman Chemical

90400-1 90400-10 90400-5
CAYM90400-1EA 3100 CZK
CAYM90400-1 CAYM90400-10 CAYM90400-5
9,12-Octadecadiynoic acid
9,12-Octadecadiynoic acid
Vzorec: C₁₈H₂₈O₂
MDL Number: MFCD00797685
CAS číslo: 2012-14-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR