Tisk…

1H,1H,2H,2H-Perfluorooctyltrichlorosilane ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

L16606.09 L16606.18
L16606.09EA 3570 CZK
L16606.09 L16606.18
1H,1H,2H,2H-Perfluorooctyltrichlorosilane ≥97%
1H,1H,2H,2H-Perfluorooctyltrichlorosilane

Purity: 97% CAS No.: 78560-45-9 Molecular Formula: CF3(CF2)5(CH2)2SiCl3 Formula Weight: 481.55 Flash Point: 87°(189°F) Refractive Index: 1.3520 Boiling Point: 192° Beilstein Reference: 6154526 UN No.: 2987 MDL No.: MFCD00042363

Vzorec: C₈H₄Cl₃F₁₃Si
MW: 481,55 g/mol
Bod varu: 192 °C
Hustota: 1,638
Bod vzplanutí: 87 °C (189 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042363
CAS číslo: 78560-45-9
EINECS: 278-947-6
UN: 2987
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR