Tisk…

Allantoin, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1-(2,5-Dioxoimidazolidin-4-yl)urea

M-Clarity™ quality level = MQ100

05670-100G 05670-25G
SIAL05670-100GEA 1030 CZK
SIAL05670-100G SIAL05670-25G
Allantoin, Sigma-Aldrich®
Allantoin
Vzorec: C₄H₆N₄O₃
MDL Number: MFCD00005260
CAS číslo: 97-59-6
EINECS: 202-592-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR