1-Boc-3-(methylamino)piperidine 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3-(Methylamino)piperidine, N1-BOC protected, 1-Boc-3-methylaminopieridine
H52773.03
H52773.06
H52773.03EA
1500
CZK
H52773.03
H52773.06
1-Boc-3-(methylamino)piperidine 97%
1-Boc-3-(methylamino)piperidine
Purity: 97%
CAS No.: 392331-89-4
Molecular Formula: C11H22N2O2
Formula Weight: 214.31
Boiling Point: 95-99°/0.6mm
MDL No.: MFCD04114980
Vzorec:
C₁₁H₂₂N₂O₂ MW: 214,31 g/mol Bod varu: 95…99 °C (0,6 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD04114980 CAS číslo: 392331-89-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...