Tisk…

(S)-(+)-3-Amino-1-boc-piperidine 97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: (S)-1-Boc-3-aminopiperidine, (S)-tert-Butyl 3-aminopiperidine-1-carboxylate, (S)-(+)-3-Amino-1-BOC-piperidine

H30538.03 H30538.06
H30538.03EA 5870 CZK
H30538.03 H30538.06
(S)-(+)-3-Amino-1-boc-piperidine 97%
(S)-(+)-3-Amino-1-boc-piperidine

Purity: 97% CAS No.: 625471-18-3 Molecular Formula: C10H20N2O2 Formula Weight: 200.28 UN No.: 2735 MDL No.: MFCD03094718

Vzorec: C₁₀H₂₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03094718
CAS číslo: 625471-18-3
UN: 2735
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR