(R)-(-)-3-Amino-1-boc-piperidine 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
tert-butyl (3R)-3-aminopiperidine-1-carboxylate, (R)-1-Boc-3-Aminopiperidine
H26937.03
H26937.06
H26937.03EA
5430
CZK
H26937.03
H26937.06
(R)-(-)-3-Amino-1-boc-piperidine 98%
(R)-(-)-3-Amino-1-boc-piperidine
Purity: 98%
CAS No.: 188111-79-7
Molecular Formula: C10H20N2O2
Formula Weight: 200.28
Physical Form: Liquid
Refractive Index: 1.4730
Sensitivity: Air Sensitive
Appearance: Colourless to pale yellow
Optical Rotation: -28° (neat)
UN No.: 2735
MDL No.: MFCD03094717
Vzorec:
C₁₀H₂₀N₂O₂ MW: 200.28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03094717 CAS číslo: 188111-79-7 UN: 2735 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...