Tisk…

(±)-3-(Boc-amino)piperidine 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: tert-Butyl piperidin-3-ylcarbamate, 3-Aminopiperidine, 3-BOC protected

H52788.03 H52788.06
H52788.03EA 1630 CZK
H52788.03 H52788.06
(±)-3-(Boc-amino)piperidine 97%
(±)-3-(Boc-amino)piperidine

Purity: 97% CAS No.: 172603-05-3 Molecular Formula: C10H20N2O2 Formula Weight: 200.28 MDL No.: MFCD03839941

Vzorec: C₁₀H₂₀N₂O₂
MW: 200,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03839941
CAS číslo: 172603-05-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR