(S)-3-(Boc-amino)piperidine 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
(S)-3-Boc-aminopiperidine, (S)-tert-Butyl piperidin-3-ylcarbamate, tert-butyl N-[(3S)-piperidin-3-yl]carbamate
H26957.03
H26957.06
H26957.03EA
2230
CZK
H26957.03
H26957.06
(S)-3-(Boc-amino)piperidine 97%
(S)-3-(Boc-amino)piperidine
Purity: 97%
CAS No.: 216854-23-8
Molecular Formula: C10H20N2O2
Formula Weight: 200.28
Melting Point: 122-127°
Optical Rotation: -15.0° (c=1 in MeOH)
Vzorec:
C₁₀H₂₀N₂O₂ Bod tání: 122…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD03093383 CAS číslo: 216854-23-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...