Tisk…

Geranyl bromide 96%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A14093.06 A14093.14 A14093.22
A14093.06EA 1840 CZK
A14093.06 A14093.14 A14093.22
Geranyl bromide 96%
Geranyl bromide

Purity: 96% CAS No.: 6138-90-5 Molecular Formula: C10H17Br Formula Weight: 217.16 Storage Temperature: 2°C to 8°C Flash Point: 95°(203°F) Refractive Index: 1.5040 Boiling Point: 99-101°/10mm Beilstein Reference: 1422412 MDL No.: MFCD00000243

Vzorec: C₁₀H₁₇Br
Bod varu: 99…101 °C (10 mmHg)
Hustota: 1,094
Bod vzplanutí: 95 °C (203 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000243
CAS číslo: 6138-90-5
EINECS: 228-123-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR