Tisk…

Ionomycin (from Streptomyces conglobatus), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

I9657-1MG I9657-5MG
SIALI9657-1MGEA 11350 CZK
SIALI9657-1MG SIALI9657-5MG
Ionomycin (from Streptomyces conglobatus), Sigma-Aldrich®
Ionomycin
Vzorec: C₄₁H₇₂O₉
MDL Number: MFCD06798385
CAS číslo: 56092-81-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR