2,3-Diphenylmaleic anhydride ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3,4-Diphenylfuran-2,5-dione
L15612.03
L15612.06
L15612.03EA
1300
CZK
L15612.03
L15612.06
2,3-Diphenylmaleic anhydride ≥98%
2,3-Diphenylmaleic anhydride
Purity: 98%
CAS No.: 4808-48-4
Molecular Formula: C16H10O3
Formula Weight: 250.26
Melting Point: 157-160°
Boiling Point: 236°/15mm
MDL No.: MFCD00005521
Vzorec:
C₁₆H₁₀O₃ MW: 250,26 g/mol Bod varu: 236 °C (15 mmHg) Bod tání: 157…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005521 CAS číslo: 4808-48-4 EINECS: 225-370-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...