Ergocalciferol (vitamin D2, calciferol), Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
E5750-1G
E5750-25G
E5750-5G
SIALE5750-1GEA
2670
CZK
SIALE5750-1G
SIALE5750-25G
SIALE5750-5G
Ergocalciferol (vitamin D2, calciferol), Sigma-Aldrich®
Ergocalciferol (vitamin D2, calciferol)
Vzorec:
C₂₈H₄₄O |
MDL Number:
MFCD00166988 CAS číslo: 50-14-6 EINECS: 200-014-9 UN: 2811 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...