Tisk…

Ergocalciferol (vitamin D2, calciferol), Sigma-Aldrich®

Dodavatel: Merck

95220-1G 95220-5G
SIAL95220-1GEA 3140 CZK
SIAL95220-1G SIAL95220-5G
Ergocalciferol (vitamin D2, calciferol), Sigma-Aldrich®
Ergocalciferol (vitamin D2, calciferol)
Vzorec: C₂₈H₄₄O
MDL Number: MFCD00166988
CAS číslo: 50-14-6
EINECS: 200-014-9
UN: 2811
ADR: 6.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR