Tisk…
2-Sulphobenzoic cyclic anhydride ≥94%
Dodavatel: Thermo Scientific
Varování
Synonyms: 2-sulphobenzoic anhydride, 2-sulphobenzoic acid cyclic anhydride
A13314.14 A13314.22 A13314.36
A13314.14EA 2440 CZK
A13314.14 A13314.22 A13314.36
2-Sulphobenzoic cyclic anhydride ≥94%
2-Sulphobenzoic cyclic anhydride
Purity: 94% CAS No.: 81-08-3 Molecular Formula: C7H4O4S Formula Weight: 184.17 Melting Point: ca 120° Boiling Point: 184-186°/18mm MDL No.: MFCD00005879
Vzorec: C₇H₄O₄S
MW: 184,17 g/mol
Bod varu: 184…186 °C (18 mmHg)
Bod tání: ∼ 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00005879
CAS číslo: 81-08-3
EINECS: 201-322-6