Tisk…

Lansoprazole, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-(((3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methyl)sulphinyl)-1H-benzo[d]imidazole

M-Clarity™ quality level = MQ100

L8533-1G L8533-250MG
SIALL8533-1GEA 11530 CZK
SIALL8533-1G SIALL8533-250MG
Lansoprazole, Sigma-Aldrich®
Lansoprazole
Vzorec: C₁₆H₁₄F₃N₃O₂S
MW: 369,36 g/mol
MDL Number: MFCD00866873
CAS číslo: 103577-45-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR