D(-)-Luciferin potassium salt
Dodavatel: Biotium
Synonyms:
(S)-2-(6-Hydroxy-2-benzothiazolyl)-2-thiazoline-4-carboxylic acid potassium salt, Beetle Luciferin potassium salt, D-Luciferin potassium, Luciferin potassium salt, Firefly luciferin potassium salt, Potassium (S)-2-(6-hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylate
10101-2
10101
10101-1
10101-2EA
33090
CZK
10101-2
10101.
10101-1
D(-)-Luciferin potassium salt
D-(-)-Luciferin potassium salt
Vzorec:
C₁₁H₇KN₂O₃S₂ |
MDL Number:
MFCD00044928 CAS číslo: 115144-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...