Retinoic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
all-trans-Retinoic acid, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid, ATRA, Vitamin A acid, Tretinoin
44540.02
44540.04
44540.77
44540.02EA
5010
CZK
44540.02
44540.04
44540.77
Retinoic acid ≥98%
Retinoic acid
Purity: 98%
CAS No.: 302-79-4
Molecular Formula: C20H28O2
Formula Weight: 300.44
Physical Form: Crystalline
Sensitivity: Light Sensitive
Beilstein Reference: 2057223
Merck Reference: 14,8165
MDL No.: MFCD00001551
Vzorec:
C₂₀H₂₈O₂ MW: 300.44 g/mol Bod tání: 180…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001551 CAS číslo: 302-79-4 EINECS: 206-129-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...