ApNA (arachidonoyl-p-nitroaniline)
Dodavatel: Cayman Chemical
10168-10
10168-5
10168-50
10168-1
CAYM10168-10EA
4450
CZK
CAYM10168-10
CAYM10168-5
CAYM10168-50
CAYM10168-1
ApNA (arachidonoyl-p-nitroaniline)
ApNA (arachidonoyl-p-nitroaniline)
Vzorec:
C₂₆H₃₆N₂O₃ |
CAS číslo:
119520-58-0 UN: 1231 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...